Bis(2,2,2-trifluoroethyl) (methoxycarbonylmethyl)phosphonate ≥95.0%
Supplier: TCI America
B1714-5G
B1714-1G
B1714-25G
TCB1714-5GEA
276.5
CAD
TCB1714-5G
TCB1714-001G
TCB1714-25G
Bis(2,2,2-trifluoroethyl) (methoxycarbonylmethyl)phosphonate ≥95.0%
Bis(2,2,2-trifluoroethyl) (methoxycarbonylmethyl)phosphonate
CAS Number: 88738-78-7
MDL Number: MFCD00009901
Molecular Formula: C7H9F6O5P
Molecular Weight: 318.11
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.50
Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: Bis(2,2,2-trifluoroethyl)phosphonoacetic Acid Methyl Ester, Methyl [Bis(2,2,2-trifluoroethoxy)phosphinyl]acetate, Methyl Bis(2,2,2-trifluoroethyl)phosphonoacetate
Formula:
C₇H₉F₆O₅P Density: 1.50 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00009901 CAS Number: 88738-78-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...