2,2'-Bis(trifluoromethyl)benzidine ≥98.0%
Supplier: TCI America
Synonyms:
2,2'-Bis(trifluoromethyl)-[1,1'-biphenyl]-4,4'-diamine, 4,4'-Diamino-2,2'-bis(trifluoromethyl)biphenyl
CAS Number: 341-58-2
MDL Number: MFCD00190155
Molecular Formula: C14H10F6N2
Molecular Weight: 320.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 183
Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials, Biphenyls (for High-Performance Polymer Research)
Other Product Names / Synonyms: 4,4'-Diamino-2,2'-bis(trifluoromethyl)biphenyl
Formula:
C₁₄H₁₀F₆N₂ MW: 320.24 g/mol Melting Pt: 183 °C |
MDL Number:
MFCD00190155 CAS Number: 341-58-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...