5-Bromo-2,3-dihydrobenzofuran-7-carboxylic acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 5-Bromocoumaran-7-carboxylic Acid

B1705-5G B1705-1G
TCB1705-005GEA 821.6 CAD
TCB1705-005G TCB1705-001G
5-Bromo-2,3-dihydrobenzofuran-7-carboxylic acid ≥97.0% (by titrimetric analysis)
5-Bromo-2,3-dihydrobenzofuran-7-carboxylic acid

CAS Number: 41177-72-4
MDL Number: MFCD00191391
Molecular Formula: C9H7BrO3
Molecular Weight: 243.06
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 232


Other Product Names / Synonyms: 5-Bromocoumaran-7-carboxylic Acid

Formula: C₉H₇BrO₃
MW: 243.06 g/mol
Melting Pt: 232 °C
MDL Number: MFCD00191391
CAS Number: 41177-72-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d5e36e0b73bb1148b60c68e24d14fe1334f99ed0&vid=d5e36e0b73bb1148b60c68e24d14fe1334f99ed0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR