N-(tert-Butyl)hydroxylamine hydrochloride ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America

B1700-5G B1700-1G
TCB1700-005GEA 440.82 CAD
TCB1700-005G TCB1700-001G
N-(tert-Butyl)hydroxylamine hydrochloride ≥98.0% (by total nitrogen and titration analysis)
N-(tert-Butyl)hydroxylamine hydrochloride

CAS Number: 57497-39-9
MDL Number: MFCD00012598
Molecular Formula: C4H11NO
Molecular Weight: 125.60
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 185

Formula: C₄H₁₁NO
Melting Pt: 185 °C
MDL Number: MFCD00012598
CAS Number: 57497-39-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb934675ca0e191f62c670b9a9c35e27b4b12f53&vid=bb934675ca0e191f62c670b9a9c35e27b4b12f53&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR