Amylopectin hydrate (from waxy corn) amylose-free
Supplier: TCI America
A0456-25G
A0456-500G
TCA0456-025GEA
48.24
CAD
TCA0456-025G
TCA0456-500G
Amylopectin hydrate (from waxy corn) amylose-free
Amylopectin hydrate
CAS Number: 9037-22-3
MDL Number: MFCD00130510
Form: Crystal
Color: White
Product Categories: Materials Chemistry, Polymers, Bioscience, Polysaccharides
Formula:
(C6H10O5)n·xH2O |
MDL Number:
MFCD00130510 CAS Number: 9037-22-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...