2,2-Bis(4-oxocyclohexyl)propane ≥95.0%

Supplier: TCI America

Synonyms: 4,4'-Isopropylidenebis(cyclohexanone)

B1672-5G B1672-25G
TCB1672-5GEA 254.38 CAD
TCB1672-5G TCB1672-025G
2,2-Bis(4-oxocyclohexyl)propane ≥95.0%
2,2-Bis(4-oxocyclohexyl)propane

CAS Number: 7418-16-8
MDL Number: MFCD00191618
Molecular Formula: C15H24O2
Molecular Weight: 236.36
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 164


Other Product Names / Synonyms: 4,4'-Isopropylidenebis(cyclohexanone)

Formula: C₁₅H₂₄O₂
MW: 236.36 g/mol
Melting Pt: 164 °C
MDL Number: MFCD00191618
CAS Number: 7418-16-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=439d4ba0b94783a52ac16716663a758906026cb4&vid=439d4ba0b94783a52ac16716663a758906026cb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR