4,4'-Bis(4-aminophenoxy)biphenyl ≥98.0%

Supplier: TCI America

Synonyms: 4,4-Bis(4-aminophenoxy)biphenyl, 4,4′-Bis(4-aminophenoxy)biphenyl

B1671-25G B1671-5G
TCB1671-25GEA 265.28 CAD
TCB1671-25G TCB1671-5G
4,4'-Bis(4-aminophenoxy)biphenyl ≥98.0%
4,4'-Bis(4-aminophenoxy)biphenyl

CAS Number: 13080-85-8
MDL Number: MFCD00039151
Molecular Formula: C24H20N2O2
Molecular Weight: 368.44
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 199

Formula: C₂₄H₂₀N₂O₂
MW: 368.44 g/mol
Melting Pt: 199 °C
MDL Number: MFCD00039151
CAS Number: 13080-85-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b50667c259766d88948befa94b91c4ab8657a1a&vid=1b50667c259766d88948befa94b91c4ab8657a1a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR