Bis(4-methacryloylthiophenyl)sulfide ≥97.0% (by HPLC)

Supplier: TCI America

Synonyms: 4,4'-Thiodibenzenedithiol Dimethacrylate

B1662-25G B1662-5G
TCB1662-025GEA 2102.98 CAD
TCB1662-025G TCB1662-5G
Bis(4-methacryloylthiophenyl)sulfide ≥97.0% (by HPLC)
Bis(4-methacryloylthiophenyl)sulfide

CAS Number: 129283-82-5
MDL Number: MFCD00142120
Molecular Formula: C20H18O2S3
Molecular Weight: 386.54
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Melting point (°C): 64
Flash Point (°C): 160
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Monomers, Diphenyl Sulfides (for High-Performance Polymer Research)
Other Product Names / Synonyms: 4,4'-Thiodibenzenedithiol Dimethacrylate

Formula: C₂₀H₁₈O₂S₃
MW: 386.54 g/mol
Melting Pt: 64 °C
Flash Pt: 160 °C
MDL Number: MFCD00142120
CAS Number: 129283-82-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6fff77373902c37a88300fb31bd8ace52fb6e15d&vid=6fff77373902c37a88300fb31bd8ace52fb6e15d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR