3-Bromo-1-(trimethylsilyl)-1-propyne ≥97.0%
Supplier: TCI America
B1654-1G
B1654-5G
B1654-25G
TCB1654-001GEA
55.3
CAD
TCB1654-001G
TCB1654-005G
TCB1654-25G
3-Bromo-1-(trimethylsilyl)-1-propyne ≥97.0%
3-Bromo-1-(trimethylsilyl)-1-propyne
CAS Number: 38002-45-8
MDL Number: MFCD00134460
Molecular Formula: C6H11BrSi
Molecular Weight: 191.14
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 62
Specific Gravity (20/20): 1.18
Product Categories: Synthetic Organic Chemistry, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: (3-Bromo-1-propynyl)trimethylsilane
Formula:
C₆H₁₁BrSi Density: 1.18 (20 °C) Flash Pt: 62 °C |
MDL Number:
MFCD00134460 CAS Number: 38002-45-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...