1,1-Bis(methylthio)ethylene ≥90.0%

Supplier: TCI America

B1650-5G B1650-1G
TCB1650-005GEA 952.74 CAD
TCB1650-005G TCB1650-001G
1,1-Bis(methylthio)ethylene ≥90.0%
1,1-Bis(methylthio)ethylene

CAS Number: 51102-74-0
MDL Number: MFCD00191617
Molecular Formula: C4H8S2
Molecular Weight: 120.23
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 110
Specific Gravity (20/20): 1.07
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Sulfides
Other Product Names / Synonyms: Ketene Dimethyl Thioacetal

Formula: C₄H₈S₂
Boiling Pt: 110 °C
Density: 1.07 (20 °C)
MDL Number: MFCD00191617
CAS Number: 51102-74-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9d9ba33c27b8cb100373a76931917ba9b82c49f&vid=b9d9ba33c27b8cb100373a76931917ba9b82c49f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR