1-Bromo-2-naphthaldehyde ≥96.0%
Supplier: TCI America
B1647-5G
B1647-1G
B1647-25G
TCB1647-5GEA
412.38
CAD
TCB1647-5G
TCB1647-001G
TCB1647-25G
1-Bromo-2-naphthaldehyde ≥96.0%
1-Bromo-2-naphthaldehyde
CAS Number: 3378-82-3
MDL Number: MFCD00046368
Molecular Formula: C11H7BrO
Molecular Weight: 235.08
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 118
Formula:
C₁₁H₇BrO MW: 235.0767 g/mol Melting Pt: 118 °C |
MDL Number:
MFCD00046368 CAS Number: 3378-82-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...