Bis(2-formylphenyl)ether ≥98.0%

Supplier: TCI America

B1646-1G B1646-10G
TCB1646-001GEA 145.36 CAD
TCB1646-001G TCB1646-010G
Bis(2-formylphenyl)ether ≥98.0%
Bis(2-formylphenyl)ether

CAS Number: 49590-51-4
MDL Number: MFCD00191616
Molecular Formula: C14H10O3
Molecular Weight: 226.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 182
Melting point (°C): 78


Product Categories: Materials Chemistry, Diphenyl Ethers (for High-Performance Polymer Research)
Other Product Names / Synonyms: 2,2'-Diformyldiphenyl Ether, 2,2'-Oxydibenzaldehyde

Formula: C₁₄H₁₀O₃
Boiling Pt: 182 °C
Melting Pt: 78 °C
MDL Number: MFCD00191616
CAS Number: 49590-51-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f8fa8b2e904b4fd2b08c6a75804b659770923c9&vid=6f8fa8b2e904b4fd2b08c6a75804b659770923c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR