Tris(hexafluoroisopropyl)borate ≥95.0% (by GC, titration analysis)
Supplier: TCI America
B1645-5G
B1645-1G
TCB1645-5GEA
826.34
CAD
TCB1645-5G
TCB1645-001G
Tris(hexafluoroisopropyl)borate ≥95.0% (by GC, titration analysis)
Tris(hexafluoroisopropyl)borate
CAS Number: 6919-80-8
MDL Number: MFCD00191615
Molecular Formula: C9H3BF18O3
Molecular Weight: 511.90
Purity/Analysis Method: >95.0% (GC,T)
Form: Crystal
Melting point (°C): 31
Other Product Names / Synonyms: Boric Acid Tris(hexafluoroisopropyl) Ester
Formula:
C₉H₃BF₁₈O₃ Melting Pt: 31 °C |
MDL Number:
MFCD00191615 CAS Number: 6919-80-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...