2,2'-Bis(hydroxymethyl)diphenyl ether ≥98.0%

Supplier: TCI America

B1643-5G B1643-1G
TCB1643-5GEA 904.08 CAD
TCB1643-5G TCB1643-001G
2,2'-Bis(hydroxymethyl)diphenyl ether ≥98.0%
2,2'-Bis(hydroxymethyl)diphenyl ether

CAS Number: 10038-40-1
MDL Number: MFCD00191613
Molecular Formula: C14H14O3
Molecular Weight: 230.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 99


Product Categories: Materials Chemistry, Monomers, Diphenyl Ethers (for High-Performance Polymer Research)
Other Product Names / Synonyms: 2,2'-Oxydi(benzyl Alcohol)

Formula: C₁₄H₁₄O₃
Melting Pt: 99 °C
MDL Number: MFCD00191613
CAS Number: 10038-40-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bcf5cb9af6650b00681d7797a15d78cf8c28f9a0&vid=bcf5cb9af6650b00681d7797a15d78cf8c28f9a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR