Nɑ-(tert-Butoxycarbonyl)-N-ε-carbobenzoxy-L-lysine ≥98.0%
Supplier: TCI America
Synonyms:
Nepsilon-Benzyloxycarbonyl-Nα-Boc-L-lysine, Boc-Lys(Z)-OH, Nɑ-Boc-Nepsilon-Cbz-L-lysine, Boc-Lys(Cbz)-OH
CAS Number: 2389-45-9
MDL Number: MFCD00065584
Molecular Formula: C19H28N2O6
Molecular Weight: 380.44
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 77
Specific rotation [a]20/D: -8 deg (C=2.5, AcOH)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nalpha-Boc-Nepsilon-Cbz-L-lysine, Boc-Lys(Z)-OH
Formula:
C₁₉H₂₈N₂O₆ MW: 380.44 g/mol Melting Pt: 77 °C |
MDL Number:
MFCD00065584 CAS Number: 2389-45-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...