Nα-(tert-Butoxycarbonyl)-L-arginine hydrochloride monohydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
B1626-5G
B1626-25G
TCB1626-005GEA
55.3
CAD
TCB1626-005G
TCB1626-25G
Nα-(tert-Butoxycarbonyl)-L-arginine hydrochloride monohydrate ≥98.0% (by titrimetric analysis)
Nalpha-Boc-L-arginine hydrochloride monohydrate
CAS Number: 35897-34-8
MDL Number: MFCD00063594
Molecular Formula: C11H22N4O4
Molecular Weight: 310.78
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: -8 deg (C=2, H2O)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nalpha-Boc-L-arginine Hydrochloride, Boc-Arg-OH.HCl
Formula:
C₁₁H₂₂N₄O₄·HCl·H₂O |
MDL Number:
MFCD00063594 CAS Number: 35897-34-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...