1,3-Bis(2-hydroxyethoxy)benzene ≥98.0%

Supplier: TCI America

B1625-500G B1625-25G
TCB1625-500GEA 643.06 CAD
TCB1625-500G TCB1625-025G
1,3-Bis(2-hydroxyethoxy)benzene ≥98.0%
1,3-Bis(2-hydroxyethoxy)benzene

CAS Number: 102-40-9
MDL Number: MFCD00016566
Molecular Formula: C10H14O4
Molecular Weight: 198.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 234
Melting point (°C): 90


Other Product Names / Synonyms: Resorcinol Bis(beta-hydroxyethyl) Ether

Formula: C₁₀H₁₄O₄
Boiling Pt: 234 °C
Melting Pt: 90 °C
MDL Number: MFCD00016566
CAS Number: 102-40-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c52c21e0cf4561f9c53bf5daa9820970d4703d2e&vid=c52c21e0cf4561f9c53bf5daa9820970d4703d2e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR