1-Bromo-2-chloro-5-nitrobenzene ≥97.0%
Supplier: TCI America
B1624-5G
B1624-25G
TCB1624-5GEA
61.62
CAD
TCB1624-5G
TCB1624-025G
1-Bromo-2-chloro-5-nitrobenzene ≥97.0%
1-Bromo-2-chloro-5-nitrobenzene
CAS Number: 16588-26-4
MDL Number: MFCD00100437
Molecular Formula: C6H3BrClNO2
Molecular Weight: 236.45
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 58
Formula:
C₆H₃BrClNO₂ Melting Pt: 58 °C |
MDL Number:
MFCD00100437 CAS Number: 16588-26-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...