1-(3-Bromopropyl)pyrrole ≥97.0%

Supplier: TCI America

B1623-1G B1623-5G
TCB1623-001GEA 380.15 CAD
TCB1623-001G TCB1623-005G
1-(3-Bromopropyl)pyrrole ≥97.0%
1-(3-Bromopropyl)pyrrole

CAS Number: 100779-91-7
MDL Number: MFCD00191324
Molecular Formula: C7H10BrN
Molecular Weight: 188.07
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 78
Specific Gravity (20/20): 1.36
Storage Temperature: 0-10°C

Formula: C₇H₁₀BrN
Boiling Pt: 78 °C
Density: 1.36 (20 °C)
MDL Number: MFCD00191324
CAS Number: 100779-91-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1db5b98ad7657ac9b4590548cce74a3a065df5c&vid=b1db5b98ad7657ac9b4590548cce74a3a065df5c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR