1-(2-Bromoethyl)pyrrole ≥97.0%
Supplier: TCI America
B1622-1G
B1622-5G
TCB1622-001GEA
214.88
CAD
TCB1622-001G
TCB1622-005G
1-(2-Bromoethyl)pyrrole ≥97.0%
1-(2-Bromoethyl)pyrrole
CAS Number: 78358-86-8
MDL Number: MFCD00191322
Molecular Formula: C6H8BrN
Molecular Weight: 174.04
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.46
Storage Temperature: 0-10°C
Formula:
C₆H₈BrN Density: 1.46 (20 °C) |
MDL Number:
MFCD00191322 CAS Number: 78358-86-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...