4,4'-Bicyclohexanol ≥98.0%
Supplier: TCI America
Synonyms:
1,1'-Bicyclohexyl-4,4'-diol, [1,1'-Bi(cyclohexane)]-4,4'-diol, 4,4'-Dihydroxydicyclohexyl
B1621-500G
B1621-25G
TCB1621-500GEA
1023.21
CAD
TCB1621-500G
TCB1621-025G
4,4'-Bicyclohexanol ≥98.0%
4,4'-Bicyclohexanol
CAS Number: 20601-38-1
MDL Number: MFCD00012056
Molecular Formula: C12H22O2
Molecular Weight: 198.31
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,1'-Bicyclohexyl-4,4'-diol, 4,4'-Dihydroxydicyclohexyl
Formula:
C₁₂H₂₂O₂ MW: 198.31 g/mol |
MDL Number:
MFCD00012056 CAS Number: 20601-38-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...