1-Bromo-2-naphthoic acid ≥98.0%

Supplier: TCI America

Synonyms: 1-Bromonaphthalene-2-carboxylic acid

B1617-100MG B1617-5G B1617-1G
TCB1617-100MGEA 75.84 CAD
TCB1617-100MG TCB1617-5G TCB1617-001G
1-Bromo-2-naphthoic acid ≥98.0%
1-Bromo-2-naphthoic acid

CAS Number: 20717-79-7
MDL Number: MFCD00021408
Molecular Formula: C11H7BrO2
Molecular Weight: 251.08
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 191


Other Product Names / Synonyms: 1-Bromo-2-naphthalenecarboxylic Acid

Formula: C₁₁H₇BrO₂
Melting Pt: 191 °C
MDL Number: MFCD00021408
CAS Number: 20717-79-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e167f2efe987bb28142a54437442cce8cca27d90&vid=e167f2efe987bb28142a54437442cce8cca27d90&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR