(-)-4,5-Bis[hydroxy(diphenyl)methyl]-2,2-dimethyl-1,3-dioxolane ≥97.0%

Supplier: TCI America

Synonyms: (4R,5R)-2,2-Dimethyl-alpha,alpha,alpha',alpha'-tetraphenyldioxolane-4,5-dimethanol, (R,R)-TADDOL, (4R,5R)-(-)-2,2-Dimethyl-α,α,α',α'-tetraphenyl-1,3-dioxolane-4,5-dimethanol, (-)-2,3-O-Isopropylidene-1,1,4,4-tetraphenyl-L-threitol

B1614-5G B1614-1G
TCB1614-005GEA 252.8 CAD
TCB1614-005G TCB1614-1G
(-)-4,5-Bis[hydroxy(diphenyl)methyl]-2,2-dimethyl-1,3-dioxolane ≥97.0%
(-)-4,5-Bis[hydroxy(diphenyl)methyl]-2,2-dimethyl-1,3-dioxolane

CAS Number: 93379-48-7
MDL Number: MFCD00064467
Molecular Formula: C31H30O4
Molecular Weight: 466.58
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Melting point (°C): 196
Specific rotation [a]20/D: -69 deg (C=1, CHCl3)


Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts, Chiral Auxiliaries, Asymmetric Synthesis, Bioscience, Sugar Alcohols, Sugars (Others), O-Substituted Sugars
Other Product Names / Synonyms: (-)-trans-alpha,alpha'-(2,2-Dimethyl-1,3-dioxolane-4,5-diyl)bis(diphenylmethanol), (2R,3R)-2,3-O-Isopropylidene-1,1,4,4-tetraphenyl-1,2,3,4-butanetetrol, (-)-2,3-O-Isopropylidene-1,1,4,4-tetraphenyl-L-threitol, TADDOL

Formula: C₃₁H₃₀O₄
MW: 466.58 g/mol
Melting Pt: 196 °C
MDL Number: MFCD00064467
CAS Number: 93379-48-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3330487e6d8f406d2def58a8bb6678c6ca8b7ff5&vid=3330487e6d8f406d2def58a8bb6678c6ca8b7ff5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR