2-Buten-1-ylsuccinic anhydride ≥98.0%

Supplier: TCI America

Synonyms: 4-Hexene-1,2-dicarboxylic Anhydride

B1613-25G B1613-5G
TCB1613-025GEA 590.13 CAD
TCB1613-025G TCB1613-5G
2-Buten-1-ylsuccinic anhydride ≥98.0%
2-Buten-1-ylsuccinic anhydride

CAS Number: 7538-42-3
MDL Number: MFCD03423563
Molecular Formula: C8H10O3
Molecular Weight: 154.17
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Specific Gravity (20/20): 1.13


Other Product Names / Synonyms: 4-Hexene-1,2-dicarboxylic Anhydride

Formula: C₈H₁₀O₃
Density: 1.13 (20 °C)
MDL Number: MFCD03423563
CAS Number: 7538-42-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2478a9814aff8df3df208b72c2b1ff4e05792d2d&vid=2478a9814aff8df3df208b72c2b1ff4e05792d2d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR