1-Bromo-4-dodecylbenzene ≥90.0%

Supplier: TCI America

B1612-25G B1612-5G
TCB1612-25GEA 413.96 CAD
TCB1612-25G TCB1612-005G
1-Bromo-4-dodecylbenzene ≥90.0%
1-Bromo-4-dodecylbenzene

CAS Number: 126930-72-1
MDL Number: MFCD00191386
Molecular Formula: C18H29Br
Molecular Weight: 325.33
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 191
Specific Gravity (20/20): 1.08


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4-Dodecylbromobenzene

Formula: C₁₈H₂₉Br
Boiling Pt: 191 °C
Density: 1.08 (20 °C)
MDL Number: MFCD00191386
CAS Number: 126930-72-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04a3bcba9c666c78ae5b4b11bd46a5bab8f6b4bc&vid=04a3bcba9c666c78ae5b4b11bd46a5bab8f6b4bc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR