2-Bromo-1,4-benzoquinone ≥93.0%
Supplier: TCI America
B1601-1G
B1601-5G
TCB1601-001GEA
303.36
CAD
TCB1601-001G
TCB1601-5G
2-Bromo-1,4-benzoquinone ≥93.0%
2-Bromo-1,4-benzoquinone
CAS Number: 3958-82-5
MDL Number: MFCD00189377
Molecular Formula: C6H3BrO2
Molecular Weight: 186.99
Purity/Analysis Method: >93.0% (GC)
Form: Crystal
Melting point (°C): 55
Storage Temperature: 0-10°C
Product Categories: Materials Chemistry, Molecular Conductors
Other Product Names / Synonyms: 2-Bromocyclohexadiene-1,4-dione, Bromoquinone
Formula:
C₆H₃BrO₂ Melting Pt: 55 °C |
MDL Number:
MFCD00189377 CAS Number: 3958-82-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...