2-(2-Bromoethyl)-2-methyl-1,3-dioxolane ≥96.0%
Supplier: TCI America
B1598-5G
B1598-25G
TCB1598-005GEA
446.19
CAD
TCB1598-005G
TCB1598-25G
2-(2-Bromoethyl)-2-methyl-1,3-dioxolane ≥96.0%
2-(2-Bromoethyl)-2-methyl-1,3-dioxolane
CAS Number: 37865-96-6
MDL Number: MFCD00191321
Molecular Formula: C6H11BrO2
Molecular Weight: 195.06
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 74
Specific Gravity (20/20): 1.42
Storage Temperature: 0-10°C
Formula:
C₆H₁₁BrO₂ Boiling Pt: 74 °C Density: 1.42 (20 °C) |
MDL Number:
MFCD00191321 CAS Number: 37865-96-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...