1-Benzyl-3-pyrrolidinol ≥96.0%

Supplier: TCI America

B1597-5G B1597-25G
TCB1597-005GEA 41.08 CAD
TCB1597-005G TCB1597-025G
1-Benzyl-3-pyrrolidinol ≥96.0%
1-Benzyl-3-pyrrolidinol

CAS Number: 775-15-5
MDL Number: MFCD00012132
Molecular Formula: C11H15NO
Molecular Weight: 177.25
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 143
Flash Point (°C): 110
Specific Gravity (20/20): 1.07

Formula: C₁₁H₁₅NO
MW: 177.240 g/mol
Boiling Pt: 143 °C
Density: 1.07 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00012132
CAS Number: 775-15-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aeb0019b62e4dbf9aae9f0f77c7d4e8d5d9c09e0&vid=aeb0019b62e4dbf9aae9f0f77c7d4e8d5d9c09e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR