Bis[4-(2-hydroxyethoxy)phenyl]sulfone ≥97.0%

Supplier: TCI America

Synonyms: 4-(2-Hydroxyethoxy)phenyl Sulphone, 4-(2-Hydroxyethoxy)phenyl sulfone

B1590-25G B1590-500G
TCB1590-025GEA 72.68 CAD
TCB1590-025G TCB1590-500G
Bis[4-(2-hydroxyethoxy)phenyl]sulfone ≥97.0%
Bis[4-(2-hydroxyethoxy)phenyl]sulfone

CAS Number: 27205-03-4
MDL Number: MFCD00130282
Molecular Formula: C16H18O6S
Molecular Weight: 338.37
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 181


Product Categories: Materials Chemistry, Monomers, Diphenyl Sulfones (for High-Performance Polymer Research)
Other Product Names / Synonyms: 4-(2-Hydroxyethoxy)phenyl Sulfone

Formula: C₁₆H₁₈O₆S
MW: 338.37 g/mol
Melting Pt: 181 °C
MDL Number: MFCD00130282
CAS Number: 27205-03-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4028ee4cc427aa2a7d3d9b54af1c9ae939f28b0a&vid=4028ee4cc427aa2a7d3d9b54af1c9ae939f28b0a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR