trans-1,2-Bis(phenylsulfonyl)ethylene ≥98.0% (by HPLC)
Supplier: TCI America
B1574-5G
B1574-1G
TCB1574-5GEA
222.78
CAD
TCB1574-5G
TCB1574-001G
trans-1,2-Bis(phenylsulfonyl)ethylene ≥98.0% (by HPLC)
trans-1,2-Bis(phenylsulfonyl)ethylene
CAS Number: 963-16-6
MDL Number: MFCD00066528
Molecular Formula: C14H12O4S2
Molecular Weight: 308.37
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 230
Formula:
C₁₄H₁₂O₄S₂ MW: 308.37 g/mol Melting Pt: 230 °C |
MDL Number:
MFCD00066528 CAS Number: 963-16-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...