cis-1,2-Bis(phenylsulfonyl)ethylene ≥95.0% (by HPLC)

Supplier: TCI America

B1573-25G B1573-5G B1573-1G
TCB1573-25GEA 692.51 CAD
TCB1573-25G TCB1573-005G TCB1573-001G
cis-1,2-Bis(phenylsulfonyl)ethylene ≥95.0% (by HPLC)
cis-1,2-Bis(phenylsulfonyl)ethylene

CAS Number: 963-15-5
MDL Number: MFCD00010140
Molecular Formula: C14H12O4S2
Molecular Weight: 308.37
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Melting point (°C): 101

Formula: C₁₄H₁₂O₄S₂
Melting Pt: 101 °C
MDL Number: MFCD00010140
CAS Number: 963-15-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0cee99b0bbdfc90e23621254d7e93e52f6d6c74e&vid=0cee99b0bbdfc90e23621254d7e93e52f6d6c74e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR