2,2-Bis(4-hydroxy-3-methylphenyl)propane ≥98.0%
Supplier: TCI America
Synonyms:
4,4'-(Propane-2,2-diyl)bis(2-methylphenol), 4,4'-Isopropylidenedi-o-cresol
CAS Number: 79-97-0
MDL Number: MFCD00002232
Molecular Formula: C17H20O2
Molecular Weight: 256.35
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 139
Flash Point (°C): 210
Product Categories: Materials Chemistry, Monomers, Bisphenol A type Compounds (for High-Performance Polymer Research), Pressure & Heat Sensitive Recording Materials
Other Product Names / Synonyms: 4,4'-Isopropylidenedi-o-cresol
Formula:
C₁₇H₂₀O₂ MW: 256.35 g/mol Melting Pt: 139 °C Flash Pt: 210 °C |
MDL Number:
MFCD00002232 CAS Number: 79-97-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...