ɑ,ɑ'-Bis(4-hydroxyphenyl)-1,4-diisopropylbenzene ≥98.0%
Supplier: TCI America
Synonyms:
4,4'-(1,4-Phenylenediisopropylidene)bisphenol, 1,4-Bis[2-(4-hydroxyphenyl)-2-propyl]benzene, ['1,4-Bis[2-(4-hydroxyphenyl)-2-propyl]benzene', 'Bisphenol P', "4,4'-(1,4-Phenylenediisopropylidene)bisphenol"], Bisphenol P, α,α'-Bis(4-hydroxyphenyl)-1,4-diisopropylbenzene
CAS Number: 2167-51-3
MDL Number: MFCD00191315
Molecular Formula: C24H26O2
Molecular Weight: 346.47
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 195
Product Categories: Materials Chemistry, Monomers, Bisphenol A type Compounds (for High-Performance Polymer Research), Pressure & Heat Sensitive Recording Materials
Other Product Names / Synonyms: 1,4-Bis[2-(4-hydroxyphenyl)-2-propyl]benzene, 4,4'-(1,4-Phenylenediisopropylidene)bisphenol, Bisphenol P
Formula:
C₂₄H₂₆O₂ MW: 346.47 g/mol Melting Pt: 195 °C |
MDL Number:
MFCD00191315 CAS Number: 2167-51-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...