ɑ,ɑ'-Bis(4-aminophenyl)-1,4-diisopropylbenzene ≥99.0%

Supplier: TCI America

B1562-25G B1562-500G
TCB1562-025GEA 93.22 CAD
TCB1562-025G TCB1562-500G
ɑ,ɑ'-Bis(4-aminophenyl)-1,4-diisopropylbenzene ≥99.0%
alpha,alpha'-Bis(4-aminophenyl)-1,4-diisopropylbenzene

CAS Number: 2716-10-1
MDL Number: MFCD00191313
Molecular Formula: C24H28N2
Molecular Weight: 344.50
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 165
Flash Point (°C): 252


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials, Bisphenol A type Compounds (for High-Performance Polymer Research)
Other Product Names / Synonyms: 1,4-Bis[2-(4-aminophenyl)-2-propyl]benzene, 4,4'-(1,4-Phenylenediisopropylidene)bisaniline, Bisaniline P

Formula: C₂₄H₂₈N₂
Melting Pt: 165 °C
Flash Pt: 252 °C
MDL Number: MFCD00191313
CAS Number: 2716-10-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1032457a2ba7fd51216d4cf6216aad8b060f0ff9&vid=1032457a2ba7fd51216d4cf6216aad8b060f0ff9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR