2,2-Bis[4-(4-aminophenoxy)phenyl]propane ≥98.0%
Supplier: TCI America
Synonyms:
4,4'-Isopropylidenebis[(4-aminophenoxy)benzene], 4,4'-((Propane-2,2-diylbis(4,1-phenylene))bis(oxy))dianiline
CAS Number: 13080-86-9
MDL Number: MFCD00039152
Molecular Formula: C27H26N2O2
Molecular Weight: 410.52
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 129
Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials, Bisphenol A type Compounds (for High-Performance Polymer Research)
Other Product Names / Synonyms: 4,4'-Isopropylidenebis[(4-aminophenoxy)benzene]
Formula:
C₂₇H₂₆N₂O₂ MW: 410.52 g/mol Melting Pt: 129 °C |
MDL Number:
MFCD00039152 CAS Number: 13080-86-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...