9,9-Bis(4-aminophenyl)fluorene ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B1549-25G B1549-100G
TCB1549-025GEA 175.38 CAD
TCB1549-025G TCB1549-100G
9,9-Bis(4-aminophenyl)fluorene ≥98.0% (by titrimetric analysis)
9,9-Bis(4-aminophenyl)fluorene

CAS Number: 15499-84-0
MDL Number: MFCD00039156
Molecular Formula: C25H20N2
Molecular Weight: 348.45
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 236
Flash Point (°C): 200


Product Categories: Materials Chemistry, Conducting Polymer Research Reagents, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: 4,4'-(9-Fluorenylidene)dianiline

Formula: C₂₅H₂₀N₂
Melting Pt: 236 °C
Flash Pt: 200 °C
MDL Number: MFCD00039156
CAS Number: 15499-84-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd3ee9bab39773f759cf9bef81babecca478313d&vid=cd3ee9bab39773f759cf9bef81babecca478313d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR