2-Bromo-5-methylthiophene ≥98.0% stabilized

Supplier: TCI America

Synonyms: 5-Methyl-2-bromothiophene, 2-Bromo-5-methylthiophenol

B1540-25G B1540-5G
TCB1540-25GEA 188.02 CAD
TCB1540-25G TCB1540-005G
2-Bromo-5-methylthiophene ≥98.0% stabilized
2-Bromo-5-methylthiophene

(stabilized with Copper chip + NaHCO3)
CAS Number: 765-58-2
MDL Number: MFCD00130103
Molecular Formula: C5H5BrS
Molecular Weight: 177.06
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 175
Flash Point (°C): 78
Specific Gravity (20/20): 1.57
Storage Temperature: 0-10°C

Formula: C₅H₅BrS
MW: 177.06 g/mol
Boiling Pt: 175 °C
Melting Pt: 58 °C
Density: 1.57 (20 °C)
Flash Pt: 78 °C
MDL Number: MFCD00130103
CAS Number: 765-58-2
EINECS: 212-151-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f89da32c48c2067bebda122bb43d2eaaef1c5cc9&vid=f89da32c48c2067bebda122bb43d2eaaef1c5cc9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR