2,3-Bis(2,4,5-trimethyl-3-thienyl)maleic anhydride ≥97.0%

Supplier: TCI America

B1534-100MG B1534-1G
TCB1534-100MGEA 148.52 CAD
TCB1534-100MG TCB1534-1G
2,3-Bis(2,4,5-trimethyl-3-thienyl)maleic anhydride ≥97.0%
2,3-Bis(2,4,5-trimethyl-3-thienyl)maleic anhydride

CAS Number: 112440-47-8
MDL Number: MFCD00142789
Molecular Formula: C18H18O3S2
Molecular Weight: 346.46
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 168

Formula: C₁₈H₁₈O₃S₂
MW: 346.46 g/mol
Melting Pt: 168 °C
MDL Number: MFCD00142789
CAS Number: 112440-47-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df8583cf31195d094d5b71c48a6f6cbbb152a5ba&vid=df8583cf31195d094d5b71c48a6f6cbbb152a5ba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR