(R)-1-Benzyl-3-pyrrolidinol ≥96.0%

Supplier: TCI America

Synonyms: (R)-1-Benzyl-3-hydroxypyrrolidine, (R)-(+)-1-Benzyl-3-pyrrolidinol, (R)-3-Hydroxy-1-benzylpyrrolidine

B1532-1G B1532-5G
TCB1532-1GEA 37.92 CAD
TCB1532-1G TCB1532-5G
(R)-1-Benzyl-3-pyrrolidinol ≥96.0%
(R)-1-Benzyl-3-pyrrolidinol

CAS Number: 101930-07-8
MDL Number: MFCD00075484
Molecular Formula: C11H15NO
Molecular Weight: 177.25
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 116
Flash Point (°C): 110
Specific Gravity (20/20): 1.07
Specific rotation [a]20/D: -4.6 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-1-Benzyl-3-hydroxypyrrolidine

Formula: C₁₁H₁₅NO
MW: 177.25 g/mol
Boiling Pt: 116 °C
Density: 1.07 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00075484
CAS Number: 101930-07-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=582e71a5c83fd00daff0149e424981f85fccd7da&vid=582e71a5c83fd00daff0149e424981f85fccd7da&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR