Behenic anhydride ≥97.0%

Supplier: TCI America

B1531-1G B1531-5G
TCB1531-001GEA 148.52 CAD
TCB1531-001G TCB1531-005G
Behenic anhydride ≥97.0%
Behenic anhydride

CAS Number: 55726-23-3
MDL Number: MFCD00010452
Molecular Formula: C44H86O3
Molecular Weight: 663.17
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 84


Other Product Names / Synonyms: Docosanoic Anhydride

Formula: C₄₄H₈₆O₃
Melting Pt: 84 °C
MDL Number: MFCD00010452
CAS Number: 55726-23-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=157c9fc43dcede113d52406b57b045fc979bca4e&vid=157c9fc43dcede113d52406b57b045fc979bca4e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR