D-Amygdalin ≥97.0% (by HPLC)
Supplier: TCI America
Synonyms:
Laetrile, Amygdalin, Amygdaloside, D-Mandelonitrile 6-O-β-D-glucosido-β-D-glucoside, (R)-2-Phenyl-2-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)acetonitrile, D-Mandelonitrile-β-gentiobioside
CAS Number: 29883-15-6
MDL Number: MFCD00006598
Molecular Formula: C20H27NO11
Molecular Weight: 457.43
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 213
Specific rotation [a]20/D: -40 deg (C=2, H2O)
Formula:
C₂₀H₂₇NO₁₁ MW: 457.43 g/mol Melting Pt: 213 °C |
MDL Number:
MFCD00006598 CAS Number: 29883-15-6 |
|
Specification Test Results
Purity(HPLC) | min. 97.0 area% |
Purity(with Total Nitrogen) | min. 93.0 % |
Specific rotation [a]20/D | -35.0 to -45.0 deg(C=2, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...