1,4-Bis(methoxymethyl)benzene ≥99.0%
Supplier: TCI America
Synonyms:
p-Xylene Glycol Dimethyl Ether
B1530-25G
B1530-500G
TCB1530-025GEA
47.4
CAD
TCB1530-025G
TCB1530-500G
1,4-Bis(methoxymethyl)benzene ≥99.0%
1,4-Bis(methoxymethyl)benzene
CAS Number: 6770-38-3
MDL Number: MFCD00025903
Molecular Formula: C10H14O2
Molecular Weight: 166.22
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 235
Flash Point (°C): 118
Specific Gravity (20/20): 1.01
Other Product Names / Synonyms: p-Xylene Glycol Dimethyl Ether
Formula:
C₁₀H₁₄O₂ MW: 166.22 g/mol Boiling Pt: 235 °C Density: 1.01 (20 °C) Flash Pt: 118 °C |
MDL Number:
MFCD00025903 CAS Number: 6770-38-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...