1,4-Benzenedimethanethiol ≥98.0%

Supplier: TCI America

Synonyms: alpha,alpha'-Dimercapto-p-xylene, 1,4-Bis(mercaptomethyl)benzene, 1,4-BDMT

B1521-5G B1521-1G
TCB1521-005GEA 654.12 CAD
TCB1521-005G TCB1521-1G
1,4-Benzenedimethanethiol ≥98.0%
1,4-Benzenedimethanethiol

CAS Number: 105-09-9
MDL Number: MFCD00004872
Molecular Formula: C8H10S2
Molecular Weight: 170.29
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 156
Melting point (°C): 43
Flash Point (°C): 110


Other Product Names / Synonyms: alpha,alpha'-Dimercapto-p-xylene, 1,4-Bis(mercaptomethyl)benzene

Formula: C₈H₁₀S₂
MW: 170.29 g/mol
Boiling Pt: 156 °C
Melting Pt: 43 °C
Flash Pt: 110 °C
MDL Number: MFCD00004872
CAS Number: 105-09-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=333be050f764840c69881ff448109a85ec89015e&vid=333be050f764840c69881ff448109a85ec89015e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR