Bis(1,3-propanediamine)copper(II) dichloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B1513-1G B1513-10G
TCB1513-001GEA 60.04 CAD
TCB1513-001G TCB1513-010G
Bis(1,3-propanediamine)copper(II) dichloride ≥98.0% (by titrimetric analysis)
Bis(1,3-propanediamine)copper(II) dichloride

CAS Number: 32270-93-2
MDL Number: MFCD00191318
Molecular Formula: C6H20Cl2CuN4
Molecular Weight: 282.70
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₆H₂₀Cl₂CuN₄
MW: 282.70 g/mol
MDL Number: MFCD00191318
CAS Number: 32270-93-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b25b1d341cd464dba2aa5a9caf93157ce12ed430&vid=b25b1d341cd464dba2aa5a9caf93157ce12ed430&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR