1,3-Bis(4,5-dihydro-2-oxazolyl)benzene ≥98.0%

Supplier: TCI America

Synonyms: 2,2'-(1,3-Phenylene)bis(2-oxazoline), 1,3-Bis(4,5-dihydrooxazol-2-yl)benzene

B1511-500G B1511-25G B1511-100G
TCB1511-500GEA 385.52 CAD
TCB1511-500G TCB1511-025G 76219-346
1,3-Bis(4,5-dihydro-2-oxazolyl)benzene ≥98.0%
1,3-Bis(4,5-dihydro-2-oxazolyl)benzene

CAS Number: 34052-90-9
MDL Number: MFCD00191606
Molecular Formula: C12H12N2O2
Molecular Weight: 216.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 146
Flash Point (°C): 205


Other Product Names / Synonyms: 2,2'-(1,3-Phenylene)bis(2-oxazoline)

Formula: C₁₂H₁₂N₂O₂
MW: 216.24 g/mol
Melting Pt: 146 °C
Flash Pt: 205 °C
MDL Number: MFCD00191606
CAS Number: 34052-90-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=adc47ebf5166a48e2bc1db4a2710d832612c7a8c&vid=adc47ebf5166a48e2bc1db4a2710d832612c7a8c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR