(S)-2-(4-Aminobenzamido)pentanedioic acid ≥97.0% (by titrimetric analysis)
Supplier: TCI America
A0442-1G
A0442-5G
TCA0442-001GEA
154.84
CAD
TCA0442-001G
TCA0442-005G
(S)-2-(4-Aminobenzamido)pentanedioic acid ≥97.0% (by titrimetric analysis)
(S)-2-(4-Aminobenzamido)pentanedioic acid
CAS Number: 4271-30-1
MDL Number: MFCD00042821
Molecular Formula: C12H14N2O5
Molecular Weight: 266.25
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 173
Formula:
C₁₂H₁₄N₂O₅ Melting Pt: 173 °C |
MDL Number:
MFCD00042821 CAS Number: 4271-30-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...