6-Bromo-3-methoxysalicylaldehyde ≥99.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 6-Bromo-2-hydroxy-3-methoxybenzaldehyde

B1487-5G B1487-25G B1487-1G
TCB1487-5GEA 137.46 CAD
TCB1487-5G TCB1487-25G TCB1487-001G
6-Bromo-3-methoxysalicylaldehyde ≥99.0% (by GC, titration analysis)
6-Bromo-3-methoxysalicylaldehyde

CAS Number: 20035-41-0
MDL Number: MFCD00046152
Molecular Formula: C8H7BrO3
Molecular Weight: 231.05
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 103

Formula: C₈H₇BrO₃
MW: 231.05 g/mol
Melting Pt: 103 °C
MDL Number: MFCD00046152
CAS Number: 20035-41-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd3db804bba31c43beacaa5d3b13c4017c477ada&vid=fd3db804bba31c43beacaa5d3b13c4017c477ada&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR