1,3-Bis(4-hydroxyphenoxy)benzene ≥98.0%

Supplier: TCI America

Synonyms: Resorcinol Bis(4-hydroxyphenyl) Ether

B1484-5G B1484-1G
TCB1484-5GEA 844.67 CAD
TCB1484-5G TCB1484-001G
1,3-Bis(4-hydroxyphenoxy)benzene ≥98.0%
1,3-Bis(4-hydroxyphenoxy)benzene

CAS Number: 126716-90-3
MDL Number: MFCD00142505
Molecular Formula: C18H14O4
Molecular Weight: 294.31
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 136


Product Categories: Materials Chemistry, Monomers, Diphenyl Ethers (for High-Performance Polymer Research)
Other Product Names / Synonyms: Resorcinol Bis(4-hydroxyphenyl) Ether

Formula: C₁₈H₁₄O₄
MW: 294.31 g/mol
Melting Pt: 136 °C
MDL Number: MFCD00142505
CAS Number: 126716-90-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c077f2eb3db607d4b1ca6b835f0598d12cf2810b&vid=c077f2eb3db607d4b1ca6b835f0598d12cf2810b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR