ɑ,ɑ'-Bis(4-methylbenzyloxy)-p-xylene ≥98.0%
Supplier: TCI America
Synonyms:
p-Xylylene Glycol Bis(4-methylbenzyl) Ether, α,α'-Bis(4-methylbenzyloxy)-p-xylene
B1482-1G
B1482-5G
TCB1482-001GEA
315.68
CAD
TCB1482-001G
TCB1482-005G
ɑ,ɑ'-Bis(4-methylbenzyloxy)-p-xylene ≥98.0%
alpha,alpha'-Bis(4-methylbenzyloxy)-p-xylene
CAS Number: 136861-46-6
MDL Number: MFCD00143577
Molecular Formula: C24H26O2
Molecular Weight: 346.47
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 100
Other Product Names / Synonyms: p-Xylylene Glycol Bis(4-methylbenzyl) Ether
Formula:
C₂₄H₂₆O₂ MW: 346.47 g/mol Melting Pt: 100 °C |
MDL Number:
MFCD00143577 CAS Number: 136861-46-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...