2-Bromo-5-nitrofuran ≥98.0%

Supplier: TCI America

B1470-5G B1470-1G
TCB1470-5GEA 687.3 CAD
TCB1470-5G TCB1470-001G
2-Bromo-5-nitrofuran ≥98.0%
2-Bromo-5-nitrofuran

CAS Number: 823-73-4
MDL Number: MFCD00142879
Molecular Formula: C4H2BrNO3
Molecular Weight: 191.97
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 118
Melting point (°C): 47

Formula: C₄H₂BrNO₃
Boiling Pt: 118 °C
Melting Pt: 47 °C
MDL Number: MFCD00142879
CAS Number: 823-73-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cbc6078a986c1f1d2a928c223ab2fbb4da2a1d41&vid=cbc6078a986c1f1d2a928c223ab2fbb4da2a1d41&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR